Product Details

SNP ID
rs189735318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1345402 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAACAGAAATGTGACCGATATC[C/G]AGTGTGTTAAAGACGCCGACTATTC
Phenotype
MIM: 308385 MIM: 430000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL3RA PubMed Links

Gene Details

Gene
IL3RA
Gene Name
interleukin 3 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267713.1 538 Intron NP_001254642.1
NM_002183.3 538 Missense Mutation CAG,GAG Q51E NP_002174.1
XM_005274431.4 538 Missense Mutation CAG,GAG Q51E XP_005274488.1
XM_005274432.1 538 Missense Mutation CAG,GAG Q51E XP_005274489.1
XM_017029491.1 538 Missense Mutation CAG,GAG Q51E XP_016884980.1

View Full Product Details