Product Details
- SNP ID
-
rs189776455
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:10301670 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTCTAGCTCATGCTGGACTCTCC[A/G]GCATCTGGACAGCTGCGTGTTGGCC
- Phenotype
-
MIM: 603487
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYH13
PubMed Links
Gene Details
- Gene
- MYH13
- Gene Name
- myosin heavy chain 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003802.2 |
5791 |
Missense Mutation |
CGG,TGG |
R1901W |
NP_003793.2 |
View Full Product Details