Product Details
- SNP ID
-
rs189471056
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241190102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCCCAGGAAGAGGACAGCACCGT[C/T]CTGATCGATGTGAGCCCCCCTGAGG
- Phenotype
-
MIM: 605096
MIM: 602877
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ANO7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2302054] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANO7
- Gene Name
- anoctamin 7
- Gene
- PPP1R7
- Gene Name
- protein phosphatase 1 regulatory subunit 7
There are no transcripts associated with this gene.
View Full Product Details