Product Details

SNP ID
rs190345490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27578181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACCAAGGCCAACCCCTCAAATG[A/G]TAGAATATATTGAGTTGACTCCAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf16 PubMed Links

Gene Details

Gene
C2orf16
Gene Name
chromosome 2 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032266.3 1660 Missense Mutation ATA,GTA I537V NP_115642.3
Gene
ZNF512
Gene Name
zinc finger protein 512
There are no transcripts associated with this gene.

View Full Product Details