Product Details

SNP ID
rs190327842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:42743528 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTCTCTTCTTCTTCAAAGAAC[C/T]TTTGAGCTTCTTCCACAGGCTGTCT
Phenotype
MIM: 609592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIT2 PubMed Links

Gene Details

Gene
RIT2
Gene Name
Ras like without CAAX 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272077.1 856 UTR 3 NP_001259006.1
NM_002930.3 856 Missense Mutation NP_002921.1

View Full Product Details