Product Details

SNP ID
rs190371335
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100380944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGAATGACAGCAAGAACTCTGT[A/C]TTTAATGGCTTCATTGGTAGCGTAT
Phenotype
MIM: 191050
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
WARS PubMed Links

Gene Details

Gene
WARS
Gene Name
tryptophanyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
WDR25
Gene Name
WD repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161476.1 120 Missense Mutation TAT,TCT Y7S NP_001154948.1
NM_024515.4 120 Missense Mutation TAT,TCT Y7S NP_078791.3
XM_005268056.4 120 Missense Mutation TAT,TCT Y7S XP_005268113.1
XM_005268057.3 120 Intron XP_005268114.1
XM_005268058.4 120 Intron XP_005268115.1
XM_011537150.2 120 Missense Mutation TAT,TCT Y7S XP_011535452.1
XM_011537151.2 120 Intron XP_011535453.2
XM_017021639.1 120 Intron XP_016877128.1

View Full Product Details