Product Details
- SNP ID
-
rs190522280
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184380774 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGCCATTCGGGGTGATGCGCTG[C/T]GTGCTGTGCGCCTGCGAGGCGGTGA
- Phenotype
-
MIM: 603475
MIM: 600044
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHRD
PubMed Links
Gene Details
- Gene
- CHRD
- Gene Name
- chordin
- Gene
- THPO
- Gene Name
- thrombopoietin
There are no transcripts associated with this gene.
View Full Product Details