Product Details

SNP ID
rs190522624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:50025656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATTTCATTCTCACAGCCCCG[C/G]GAGTTGTATCCCTGATTCCTGCGGT
Phenotype
MIM: 616869
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNEP1R1 PubMed Links

Gene Details

Gene
CNEP1R1
Gene Name
CTD nuclear envelope phosphatase 1 regulatory subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281789.1 Intron NP_001268718.1
NM_153261.5 Intron NP_694993.2

View Full Product Details