Product Details

SNP ID
rs190428197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:35243287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTGGAAACCATGCCTTTCCCCT[A/G]AAAGTGGTGAGGAATGGGAAATCAT
Phenotype
MIM: 609601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF397 PubMed Links

Gene Details

Gene
ZNF397
Gene Name
zinc finger protein 397
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135178.2 706 Missense Mutation AAA,GAA K184E NP_001128650.1
NM_032347.2 706 Missense Mutation AAA,GAA K184E NP_115723.1
XM_006722558.2 706 Missense Mutation AAA,GAA K203E XP_006722621.1
XM_011526229.2 706 Missense Mutation AAA,GAA K203E XP_011524531.1
XM_011526230.2 706 Missense Mutation AAA,GAA K203E XP_011524532.1
XM_011526231.2 706 Missense Mutation AAA,GAA K184E XP_011524533.1
XM_017026042.1 706 Missense Mutation AAA,GAA K184E XP_016881531.1
Gene
ZSCAN30
Gene Name
zinc finger and SCAN domain containing 30
There are no transcripts associated with this gene.

View Full Product Details