Product Details
- SNP ID
-
rs190426453
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:62370146 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCAGCAGGGCCCACGCTGTCGTG[G/T]AGTTTGCGCACATGTTTCTTTAAGT
- Phenotype
-
MIM: 607414
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FEZF2
PubMed Links
Gene Details
- Gene
- FEZF2
- Gene Name
- FEZ family zinc finger 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018008.3 |
1612 |
Silent Mutation |
CTA,CTC |
L439L |
NP_060478.3 |
View Full Product Details