Product Details

SNP ID
rs190176169
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110512815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATAATATATGTTTTCAAGAAA[A/G]TCAGCCTTTGCAAGTTATTTTTGAC
Phenotype
MIM: 608368
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD9B PubMed Links

Gene Details

Gene
RAD9B
Gene Name
RAD9 checkpoint clamp component B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286531.1 391 Missense Mutation AAT,AGT N70S NP_001273460.1
NM_001286532.1 391 Missense Mutation AAT,AGT N70S NP_001273461.1
NM_001286533.1 391 UTR 5 NP_001273462.1
NM_001286534.1 391 UTR 5 NP_001273463.1
NM_001286535.1 391 Missense Mutation AAT,AGT N142S NP_001273464.1
NM_001286536.1 391 Intron NP_001273465.1
NM_152442.3 391 Missense Mutation AAT,AGT N142S NP_689655.3
XM_005253849.4 391 Intron XP_005253906.1
XM_011537971.2 391 Missense Mutation AAT,AGT N142S XP_011536273.1
XM_011537972.2 391 Missense Mutation AAT,AGT N118S XP_011536274.1
XM_011537974.2 391 Missense Mutation AAT,AGT N70S XP_011536276.1
XM_017018877.1 391 Missense Mutation AAT,AGT N90S XP_016874366.1
XM_017018878.1 391 Missense Mutation AAT,AGT N70S XP_016874367.1
XM_017018879.1 391 Intron XP_016874368.1
XM_017018880.1 391 Intron XP_016874369.1
XM_017018881.1 391 Intron XP_016874370.1
XM_017018882.1 391 UTR 5 XP_016874371.1

View Full Product Details