Product Details

SNP ID
rs190173301
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:15462751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGCTCCTCCAGGACCTACCG[C/T]GTGGGGCTGGGCCGGCACAACCTCT
Phenotype
MIM: 609443
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CELA2A PubMed Links

Gene Details

Gene
CELA2A
Gene Name
chymotrypsin like elastase family member 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033440.2 272 Silent Mutation CGC,CGT R82R NP_254275.1

View Full Product Details