Product Details

SNP ID
rs190167352
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:26161664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGGACGCTGCCCCGAGCAGCCCG[G/T]CCTCCACAGAGCGACCTGCGGCTTC
Phenotype
MIM: 616698
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM110D PubMed Links

Gene Details

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024869.2 560 Missense Mutation GCC,TCC A125S NP_079145.2
Gene
ZNF593
Gene Name
zinc finger protein 593
There are no transcripts associated with this gene.

View Full Product Details