Product Details

SNP ID
rs190208939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149198413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGATAGGGCACCTGTCTTCCC[C/T]GACCGCATGATGCGAGAGCCCCAGT
Phenotype
MIM: 603397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF282 PubMed Links

Gene Details

Gene
ZNF282
Gene Name
zinc finger protein 282
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303481.1 374 Silent Mutation CCC,CCT P82P NP_001290410.1
NM_003575.3 374 Silent Mutation CCC,CCT P82P NP_003566.1
XM_006716151.3 374 Silent Mutation CCC,CCT P82P XP_006716214.1

View Full Product Details