Product Details

SNP ID
rs190259239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:27067424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAAAGAGCCGGCTGACTGGCCC[C/T]GCCCACAGCACCAAGCAGACTGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf82 PubMed Links

Gene Details

Gene
C16orf82
Gene Name
chromosome 16 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145545.1 527 Silent Mutation CCC,CCT P143P NP_001139017.1

View Full Product Details