Product Details

SNP ID
rs190020175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77040157 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCAACCTCACGCTCAAGCACCT[A/G]CACTTGCTCGACTGGCTGGCTGCAC
Phenotype
MIM: 615315
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
B3GNT6 PubMed Links

Gene Details

Gene
B3GNT6
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138706.4 744 Silent Mutation CTA,CTG L202L NP_619651.3

View Full Product Details