Product Details

SNP ID
rs190048553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:112534243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCGTTAACAGCCATTTTGCCA[A/C]TAATCTTACATACAGGGAAGGATAC
Phenotype
MIM: 609606
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATG3 PubMed Links

Gene Details

Gene
ATG3
Gene Name
autophagy related 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278712.1 1324 Missense Mutation GTG,TTG V297L NP_001265641.1
NM_022488.4 1324 Intron NP_071933.2
XM_011513074.1 1324 Intron XP_011511376.1

View Full Product Details