Product Details

SNP ID
rs189993261
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:247896010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGATCATGAACCCCAGGCTCTGC[C/T]GGGGCTTGGTGTCAGTGACCTGGGG
Phenotype
MIM: 616729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2W3 PubMed Links

Gene Details

Gene
OR2W3
Gene Name
olfactory receptor family 2 subfamily W member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001957.2 424 Missense Mutation CGG,TGG R142W NP_001001957.2

View Full Product Details