Product Details

SNP ID
rs190037763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57769605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGCATATTCTGCTAGCAGGGTG[A/G]GACTGATGATTCGCCACTTGTGCTT
Phenotype
MIM: 609506 MIM: 613336 MIM: 604466 MIM: 615258
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP27B1 PubMed Links

Gene Details

Gene
CYP27B1
Gene Name
cytochrome P450 family 27 subfamily B member 1
There are no transcripts associated with this gene.

Gene
MARCH9
Gene Name
membrane associated ring-CH-type finger 9
There are no transcripts associated with this gene.

Gene
METTL1
Gene Name
methyltransferase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005371.5 699 Missense Mutation CCC,CTC P178L NP_005362.3
NM_023033.3 699 Silent Mutation TCC,TCT S116S NP_075422.3
XM_005268873.2 699 Missense Mutation CCC,CTC P135L XP_005268930.1
XM_017019305.1 699 Missense Mutation CCC,CTC P203L XP_016874794.1
Gene
METTL21B
Gene Name
methyltransferase like 21B
There are no transcripts associated with this gene.

View Full Product Details