Product Details

SNP ID
rs190661380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40482236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGGTTCCTAATGGAGACCCAA[C/T]TGCAGGGGGCTTTCCTCTGGATACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF619 PubMed Links

Gene Details

Gene
ZNF619
Gene Name
zinc finger protein 619
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145082.2 492 Missense Mutation ACT,ATT T62I NP_001138554.1
NM_001145083.1 492 Intron NP_001138555.1
NM_001145093.2 492 Intron NP_001138565.1
XM_006713112.2 492 Intron XP_006713175.1
XM_006713113.3 492 Intron XP_006713176.1
XM_011533607.1 492 Missense Mutation ACT,ATT T62I XP_011531909.1
XM_011533608.1 492 Intron XP_011531910.1
XM_017006225.1 492 Intron XP_016861714.1

View Full Product Details