Product Details

SNP ID
rs190628533
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:86560300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATGTGGAGAGAAAGGCGAATA[C/T]ATTCACTTCACCCCTGACCTTCTAC
Phenotype
MIM: 616857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA4 PubMed Links

Gene Details

Gene
CLCA4
Gene Name
chloride channel accessory 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012128.3 434 Silent Mutation TAC,TAT Y130Y NP_036260.2
XM_011541015.2 434 Silent Mutation TAC,TAT Y79Y XP_011539317.1

View Full Product Details