Product Details

SNP ID
rs190884433
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135624362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCGTGGAGCTGGGCCACCAAC[C/G]GGCCCAGGGTCTCCACCCCGAACTC
Phenotype
MIM: 613699
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLT6D1 PubMed Links

Gene Details

Gene
GLT6D1
Gene Name
glycosyltransferase 6 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182974.2 1144 Missense Mutation CCG,CGG P189R NP_892019.2
XM_011518633.1 1144 Missense Mutation CCG,CGG P225R XP_011516935.1
XM_011518634.1 1144 Missense Mutation CCG,CGG P225R XP_011516936.1
XM_011518635.1 1144 Missense Mutation CCG,CGG P225R XP_011516937.1
XM_011518636.1 1144 Missense Mutation CCG,CGG P225R XP_011516938.1
XM_011518637.1 1144 Missense Mutation CCG,CGG P189R XP_011516939.1
XM_011518638.2 1144 Missense Mutation CCG,CGG P157R XP_011516940.1
Gene
LOC102723971
Gene Name
lipocalin 1-like
There are no transcripts associated with this gene.

View Full Product Details