Product Details

SNP ID
rs191015799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63832948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGATCCCTGAGCAGTTAGGTCAGG[C/T]GAATTCCCAGCACCTGTTCCAGTTC
Phenotype
MIM: 601681 MIM: 601736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FTSJ3 PubMed Links

Gene Details

Gene
FTSJ3
Gene Name
FtsJ homolog 3
There are no transcripts associated with this gene.

Gene
PSMC5
Gene Name
proteasome 26S subunit, ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199163.1 1450 Intron NP_001186092.1
NM_002805.5 1450 Intron NP_002796.4
Gene
SMARCD2
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098426.1 1450 Missense Mutation CAC,CGC H529R NP_001091896.1
XM_005257604.2 1450 Missense Mutation CAC,CGC H454R XP_005257661.2
XM_017024967.1 1450 Missense Mutation CAC,CGC H481R XP_016880456.1

View Full Product Details