Product Details

SNP ID
rs190966689
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:155290652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTCACCACAATCACCAGGTCTC[C/T]AACACGGAGGAAGCCACGGAGCTTT
Phenotype
MIM: 609973 MIM: 609712
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCN3 PubMed Links

Gene Details

Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020897.2 1621 Intron NP_065948.1
XM_011509816.2 1621 Intron XP_011508118.1
XM_011509817.2 1621 Intron XP_011508119.1
XM_011509818.2 1621 Intron XP_011508120.1
XM_017001918.1 1621 Intron XP_016857407.1
Gene
PKLR
Gene Name
pyruvate kinase, liver and RBC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000298.5 1621 Missense Mutation AGA,GGA R549G NP_000289.1
NM_181871.3 1621 Missense Mutation AGA,GGA R518G NP_870986.1
XM_006711386.3 1621 Missense Mutation AGA,GGA R485G XP_006711449.1
XM_011509640.2 1621 Missense Mutation AGA,GGA R485G XP_011507942.1
XM_017001493.1 1621 Intron XP_016856982.1

View Full Product Details