Product Details

SNP ID
rs190982884
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2180527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGCTGGAGCGCTTGGGTGGGGG[C/T]GGCGGGGGCCCCTTCTTTCGGGGCC
Phenotype
MIM: 612184 MIM: 606692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASKIN1 PubMed Links

Gene Details

Gene
CASKIN1
Gene Name
CASK interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020764.3 2873 Silent Mutation CCA,CCG P947P NP_065815.1
Gene
TRAF7
Gene Name
TNF receptor associated factor 7
There are no transcripts associated with this gene.

View Full Product Details