Product Details

SNP ID
rs191002477
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34102599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATACCGGTCTCGAACAGTTGTTGG[C/T]TGAAGATTCATTTGATGAATCTGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EMC7 PubMed Links

Gene Details

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020154.2 527 Intron NP_064539.1
Gene
PGBD4
Gene Name
piggyBac transposable element derived 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152595.4 527 Missense Mutation GCT,GTT A23V NP_689808.2

View Full Product Details