Product Details
- SNP ID
-
rs190697600
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:97960978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTATTCCAGCGGCGAGACAGTGGC[A/C]GGGCACGTGCTGCTGGAGGCGTCCG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARRDC4
PubMed Links
Gene Details
- Gene
- ARRDC4
- Gene Name
- arrestin domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_183376.2 |
276 |
Silent Mutation |
GCA,GCC |
A39A |
NP_899232.2 |
View Full Product Details