Product Details
- SNP ID
-
rs190675907
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:21017697 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGAGCGGGACCGGTTGCCATCAA[C/T]GGATGCTGCACTGGTCAGAGAGGCT
- Phenotype
-
MIM: 605272
MIM: 616956
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6717
PubMed Links
Gene Details
- Gene
- MIR6717
- Gene Name
- microRNA 6717
There are no transcripts associated with this gene.
- Gene
- NDRG2
- Gene Name
- NDRG family member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282211.1 |
1277 |
Missense Mutation |
ATT,GTT |
I335V |
NP_001269140.1 |
NM_001282212.1 |
1277 |
Missense Mutation |
ATT,GTT |
I309V |
NP_001269141.1 |
NM_001282213.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
NP_001269142.1 |
NM_001282214.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
NP_001269143.1 |
NM_001282215.1 |
1277 |
Missense Mutation |
ATT,GTT |
I328V |
NP_001269144.1 |
NM_001282216.1 |
1277 |
Missense Mutation |
ATT,GTT |
I279V |
NP_001269145.1 |
NM_001320329.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
NP_001307258.1 |
NM_016250.2 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
NP_057334.1 |
NM_201535.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
NP_963293.1 |
NM_201536.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
NP_963294.1 |
NM_201537.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
NP_963831.1 |
NM_201538.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
NP_963832.1 |
NM_201539.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
NP_963833.1 |
NM_201540.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
NP_963834.1 |
NM_201541.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
NP_963835.1 |
XM_011536996.2 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
XP_011535298.1 |
XM_011536997.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
XP_011535299.1 |
XM_011536998.1 |
1277 |
Missense Mutation |
ATT,GTT |
I339V |
XP_011535300.1 |
XM_011536999.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
XP_011535301.1 |
XM_011537001.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
XP_011535303.1 |
XM_011537002.1 |
1277 |
Missense Mutation |
ATT,GTT |
I325V |
XP_011535304.1 |
XM_017021480.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876969.1 |
XM_017021481.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876970.1 |
XM_017021482.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876971.1 |
XM_017021483.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876972.1 |
XM_017021484.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876973.1 |
XM_017021485.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876974.1 |
XM_017021486.1 |
1277 |
Missense Mutation |
ATT,GTT |
I389V |
XP_016876975.1 |
XM_017021487.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876976.1 |
XM_017021488.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876977.1 |
XM_017021489.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876978.1 |
XM_017021490.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876979.1 |
XM_017021491.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876980.1 |
XM_017021492.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876981.1 |
XM_017021493.1 |
1277 |
Missense Mutation |
ATT,GTT |
I375V |
XP_016876982.1 |
XM_017021494.1 |
1277 |
Missense Mutation |
ATT,GTT |
I357V |
XP_016876983.1 |
XM_017021495.1 |
1277 |
Missense Mutation |
ATT,GTT |
I357V |
XP_016876984.1 |
XM_017021496.1 |
1277 |
Missense Mutation |
ATT,GTT |
I343V |
XP_016876985.1 |
XM_017021497.1 |
1277 |
Missense Mutation |
ATT,GTT |
I211V |
XP_016876986.1 |
- Gene
- TPPP2
- Gene Name
- tubulin polymerization promoting protein family member 2
There are no transcripts associated with this gene.
View Full Product Details