Product Details

SNP ID
rs190684502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128513272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTGACTCAATCTAGTGTCCCA[A/G]ACATGGGGCTTTGCCTCTCCATTTC
Phenotype
MIM: 603371
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLE1 PubMed Links

Gene Details

Gene
GLE1
Gene Name
GLE1, RNA export mediator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003722.1 Intron NP_001003722.1
NM_001499.2 Intron NP_001490.1
XM_006717059.2 Intron XP_006717122.1
XM_006717060.2 Intron XP_006717123.1
XM_011518549.1 Intron XP_011516851.1
XM_011518550.1 Intron XP_011516852.1
XM_011518551.1 Intron XP_011516853.1

View Full Product Details