Product Details

SNP ID
rs190694809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30752184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTTCACTGGGGGTGATGAGTAC[C/T]AGAAGCACTTCCTGCCCAGGGACTA
Phenotype
MIM: 604854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INMT PubMed Links

Gene Details

Gene
INMT
Gene Name
indolethylamine N-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199219.1 50 Nonsense Mutation CAG,TAG Q12* NP_001186148.1
NM_006774.4 50 Nonsense Mutation CAG,TAG Q12* NP_006765.4
Gene
INMT-FAM188B
Gene Name
INMT-FAM188B readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details