Product Details

SNP ID
rs190781000
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:27669234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCCCCTTACCTGCATCCTTACC[C/T]GCATTCTCACCTGCCTCCACCCCAC
Phenotype
MIM: 147572
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFI6 PubMed Links

Gene Details

Gene
IFI6
Gene Name
interferon alpha inducible protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002038.3 204 Intron NP_002029.3
NM_022872.2 204 Silent Mutation GCA,GCG A27A NP_075010.1
NM_022873.2 204 Silent Mutation GCA,GCG A27A NP_075011.1

View Full Product Details