Product Details

SNP ID
rs190799686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28931280 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACACCCTCAAAACGTTTGTCTA[C/T]GTTATTTTTCCATAAAGCTAAGCCT
Phenotype
MIM: 613083
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LTN1 PubMed Links

Gene Details

Gene
LTN1
Gene Name
listerin E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320766.1 5225 Missense Mutation ATA,GTA I1737V NP_001307695.1
NM_015565.2 5225 Missense Mutation ATA,GTA I1751V NP_056380.2
XM_006723987.3 5225 Intron XP_006724050.1
XM_017028316.1 5225 Missense Mutation ATA,GTA I1416V XP_016883805.1
XM_017028317.1 5225 Missense Mutation ATA,GTA I994V XP_016883806.1

View Full Product Details