Product Details

SNP ID
rs190740424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143706734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTTCTGCCGCAGCTTCAGCG[C/T]TCACGCTGTCCTCCGACTGTGGGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BREA2 PubMed Links

Gene Details

Gene
BREA2
Gene Name
breast cancer estrogen-induced apoptosis 2
There are no transcripts associated with this gene.

Gene
CCDC166
Gene Name
coiled-coil domain containing 166
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162914.1 1280 Missense Mutation AAC,AGC N427S NP_001156386.1
Gene
LOC101928160
Gene Name
uncharacterized LOC101928160
There are no transcripts associated with this gene.

Gene
MAPK15
Gene Name
mitogen-activated protein kinase 15
There are no transcripts associated with this gene.

View Full Product Details