Product Details

SNP ID
rs190767592
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46500755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACATTCACTGCATTCATAGGG[C/T]TTCTCCCCTGTGTGAGTTCTCTGGT
Phenotype
MIM: 300573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF674 PubMed Links

Gene Details

Gene
ZNF674
Gene Name
zinc finger protein 674
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039891.2 1232 Silent Mutation AAA,AAG K278K NP_001034980.1
NM_001146291.1 1232 Silent Mutation AAA,AAG K272K NP_001139763.1
NM_001190417.1 1232 Silent Mutation AAA,AAG K273K NP_001177346.1
XM_011543941.2 1232 Intron XP_011542243.1
XM_011543943.2 1232 Intron XP_011542245.1
XM_011543944.2 1232 Intron XP_011542246.1
XM_011543945.2 1232 Intron XP_011542247.1
XM_017029728.1 1232 Intron XP_016885217.1
XM_017029729.1 1232 Intron XP_016885218.1
XM_017029730.1 1232 Silent Mutation AAA,AAG K211K XP_016885219.1
XM_017029731.1 1232 Intron XP_016885220.1
XM_017029732.1 1232 Intron XP_016885221.1
XM_017029733.1 1232 Intron XP_016885222.1

View Full Product Details