Product Details

SNP ID
rs190789123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40758961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTCACTCACCTAATTTTAGGTC[A/G]CTTTGCATCTCTCTGTGCCCTTGAC
Phenotype
MIM: 611992
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS31 PubMed Links

Gene Details

Gene
MRPS31
Gene Name
mitochondrial ribosomal protein S31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005830.3 661 Nonsense Mutation CGA,TGA R196* NP_005821.2

View Full Product Details