Product Details
- SNP ID
-
rs190800927
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:71826742 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTAATTTTTGTTTGCCCTTTTC[C/T]GGGTTGAAGGTTCCTCTGCTGATGA
- Phenotype
-
MIM: 604875
MIM: 604485
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYO9A
PubMed Links
Gene Details
- Gene
- MYO9A
- Gene Name
- myosin IXA
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006901.3 |
7932 |
Silent Mutation |
CCA,CCG |
P2495P |
NP_008832.2 |
XM_006720539.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2566P |
XP_006720602.1 |
XM_011521613.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2585P |
XP_011519915.1 |
XM_011521614.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2585P |
XP_011519916.1 |
XM_011521615.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2585P |
XP_011519917.1 |
XM_011521616.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2585P |
XP_011519918.1 |
XM_011521617.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2584P |
XP_011519919.1 |
XM_011521618.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2567P |
XP_011519920.1 |
XM_011521619.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2566P |
XP_011519921.1 |
XM_011521620.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2565P |
XP_011519922.1 |
XM_011521621.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2545P |
XP_011519923.1 |
XM_011521622.2 |
7932 |
Silent Mutation |
CCA,CCG |
P2513P |
XP_011519924.1 |
XM_011521623.2 |
7932 |
Silent Mutation |
CCA,CCG |
P1684P |
XP_011519925.1 |
XM_017022228.1 |
7932 |
Silent Mutation |
CCA,CCG |
P2566P |
XP_016877717.1 |
XM_017022229.1 |
7932 |
Silent Mutation |
CCA,CCG |
P2513P |
XP_016877718.1 |
XM_017022230.1 |
7932 |
Silent Mutation |
CCA,CCG |
P2494P |
XP_016877719.1 |
- Gene
- NR2E3
- Gene Name
- nuclear receptor subfamily 2 group E member 3
There are no transcripts associated with this gene.
View Full Product Details