Product Details
- SNP ID
-
rs190851583
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:36456247 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGGCTCTTCGGTAATACAGAGG[C/G]GGGAAGGTGTACTCAATTCCCAGCC
- Phenotype
-
MIM: 138971
MIM: 611979
MIM: 608854
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CSF3R
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201522148] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CSF3R
- Gene Name
- colony stimulating factor 3 receptor
There are no transcripts associated with this gene.
- Gene
- MRPS15
- Gene Name
- mitochondrial ribosomal protein S15
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031280.3 |
740 |
Silent Mutation |
CCC,CCG |
P192P |
NP_112570.2 |
- Gene
- OSCP1
- Gene Name
- organic solute carrier partner 1
There are no transcripts associated with this gene.
View Full Product Details