Product Details

SNP ID
rs190551610
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26176390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGCGAGACATCCTGCTCATCCA[C/T]TCCTGCCGGCAGTGGACAACGGTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAREM2 PubMed Links

Gene Details

Gene
GAREM2
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168241.1 299 Silent Mutation CAC,CAT H53H NP_001161713.1
NM_001191033.1 299 Intron NP_001177962.1
XM_006711951.3 299 Silent Mutation CAC,CAT H53H XP_006712014.1
XM_011532564.2 299 Silent Mutation CAC,CAT H51H XP_011530866.1
XM_011532565.2 299 Silent Mutation CAC,CAT H51H XP_011530867.1
XM_011532566.2 299 Intron XP_011530868.1
XM_011532567.2 299 Silent Mutation CAC,CAT H53H XP_011530869.1

View Full Product Details