Product Details

SNP ID
rs190547766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6536524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGTATTGGGCGCCTGGTCACCAG[A/G]GCTGCTTTTAACTCTGGTAAAGTGG
Phenotype
MIM: 138400 MIM: 610495 MIM: 615638
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAPDH PubMed Links

Gene Details

Gene
GAPDH
Gene Name
glyceraldehyde-3-phosphate dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256799.2 340 UTR 5 NP_001243728.1
NM_001289745.1 340 Silent Mutation NP_001276674.1
NM_001289746.1 340 Intron NP_001276675.1
NM_002046.5 340 Silent Mutation NP_002037.2
Gene
IFFO1
Gene Name
intermediate filament family orphan 1
There are no transcripts associated with this gene.

Gene
NCAPD2
Gene Name
non-SMC condensin I complex subunit D2
There are no transcripts associated with this gene.

View Full Product Details