Product Details

SNP ID
rs191193482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:79904211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAGACACAAGCAGGCGGTCAGC[A/G]GTGGGGAGGGCGGCGACCTGGGCTC
Phenotype
MIM: 605696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNG2 PubMed Links

Gene Details

Gene
KCNG2
Gene Name
potassium voltage-gated channel modifier subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012283.1 Intron NP_036415.1
Gene
PQLC1
Gene Name
PQ loop repeat containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146343.1 Intron NP_001139815.1
NM_001146345.1 Intron NP_001139817.1
NM_025078.4 Intron NP_079354.2
XM_005266770.2 Intron XP_005266827.1
XM_011526188.1 Intron XP_011524490.1
XM_017025997.1 Intron XP_016881486.1
XM_017025998.1 Intron XP_016881487.1
XM_017025999.1 Intron XP_016881488.1
XM_017026000.1 Intron XP_016881489.1
XM_017026001.1 Intron XP_016881490.1
XM_017026002.1 Intron XP_016881491.1
XM_017026003.1 Intron XP_016881492.1
XM_017026004.1 Intron XP_016881493.1
XM_017026005.1 Intron XP_016881494.1

View Full Product Details