Product Details
- SNP ID
-
rs191563892
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:47802062 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGGGCCTGGGATTGGAGCTGGGA[C/T]TGAGCCTGAGCCTGGGCCTGAGCCT
- Phenotype
-
MIM: 611166
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TPRX1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77144450] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TPRX1
- Gene Name
- tetrapeptide repeat homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_198479.2 |
1020 |
Missense Mutation |
ATC,GTC |
I317V |
NP_940881.2 |
View Full Product Details