Product Details

SNP ID
rs191494496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68421792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCATCATGACTGCTTCGTTTTG[C/T]CCTTCTGATTTCCACGGCACAAGAT
Phenotype
MIM: 610008 MIM: 188830 MIM: 609224
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARSG PubMed Links

Gene Details

Gene
ARSG
Gene Name
arylsulfatase G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267727.1 3437 Intron NP_001254656.1
NM_014960.4 3437 Intron NP_055775.2
XM_005257170.3 3437 Intron XP_005257227.1
XM_005257172.3 3437 Intron XP_005257229.1
XM_006721777.3 3437 Intron XP_006721840.2
XM_006721779.3 3437 Intron XP_006721842.1
XM_011524535.2 3437 Intron XP_011522837.1
XM_011524536.2 3437 Intron XP_011522838.1
XM_011524537.1 3437 Intron XP_011522839.1
XM_011524538.2 3437 Intron XP_011522840.1
XM_011524540.2 3437 Intron XP_011522842.1
XM_011524541.2 3437 Intron XP_011522843.1
XM_011524542.2 3437 Intron XP_011522844.1
XM_011524543.2 3437 Intron XP_011522845.1
XM_011524544.2 3437 Intron XP_011522846.1
XM_011524545.2 3437 UTR 3 XP_011522847.1
XM_011524546.2 3437 Intron XP_011522848.1
XM_017024360.1 3437 Intron XP_016879849.1
XM_017024361.1 3437 Intron XP_016879850.1
XM_017024362.1 3437 Intron XP_016879851.1
XM_017024363.1 3437 Intron XP_016879852.1
XM_017024364.1 3437 Intron XP_016879853.1
XM_017024365.1 3437 Intron XP_016879854.1
XM_017024366.1 3437 Intron XP_016879855.1
XM_017024367.1 3437 Intron XP_016879856.1
XM_017024368.1 3437 UTR 3 XP_016879857.1
Gene
MIR635
Gene Name
microRNA 635
There are no transcripts associated with this gene.

Gene
PRKAR1A
Gene Name
protein kinase cAMP-dependent type I regulatory subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276289.1 3437 Intron NP_001263218.1
NM_001276290.1 3437 Intron NP_001263219.1
NM_001278433.1 3437 Intron NP_001265362.1
NM_002734.4 3437 Intron NP_002725.1
NM_212471.2 3437 Intron NP_997636.1
NM_212472.2 3437 Intron NP_997637.1
XM_011524983.2 3437 Intron XP_011523285.1
XM_011524984.2 3437 Intron XP_011523286.1
XM_011524985.2 3437 Intron XP_011523287.1
Gene
WIPI1
Gene Name
WD repeat domain, phosphoinositide interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320772.1 3437 Missense Mutation GAC,GGC D359G NP_001307701.1
NM_017983.6 3437 Missense Mutation GAC,GGC D441G NP_060453.3
XM_017024808.1 3437 Missense Mutation GAC,GGC D313G XP_016880297.1

View Full Product Details