Product Details

SNP ID
rs191545207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31201722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTGCTTGGGTTGGTGGGCTCGG[C/T]CCGCACTGCCTGGTACTGCTCATCC
Phenotype
MIM: 137070 MIM: 606838
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUS PubMed Links

Gene Details

Gene
FUS
Gene Name
FUS RNA binding protein
There are no transcripts associated with this gene.

Gene
PYCARD
Gene Name
PYD and CARD domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013258.4 480 Missense Mutation ACC,GCC T151A NP_037390.2
NM_145182.2 480 Missense Mutation ACC,GCC T132A NP_660183.1
Gene
PYCARD-AS1
Gene Name
PYCARD antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details