Product Details

SNP ID
rs191525343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:40337300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTGGGACCGAGATCAGTACGAT[A/G]GCCTAGACTCCATCAGGATCCCCAG
Phenotype
MIM: 605116
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNA9 PubMed Links

Gene Details

Gene
CHRNA9
Gene Name
cholinergic receptor nicotinic alpha 9 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017581.3 440 Missense Mutation AGC,GGC S101G NP_060051.2
Gene
LOC101060498
Gene Name
uncharacterized LOC101060498
There are no transcripts associated with this gene.

View Full Product Details