Product Details
- SNP ID
-
rs191552868
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:44703407 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACCCTGGAGCCTGCCCAGCCGAG[C/T]GAAGGGGACAACGTCACGCTGGTCG
- Phenotype
-
MIM: 614591
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEACAM16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs894189] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEACAM16
- Gene Name
- carcinoembryonic antigen related cell adhesion molecule 16
View Full Product Details