Product Details

SNP ID
rs191315480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:114282095 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGCTGCGCCCGCTGCCAGCACA[C/T]GACTACTTCGAGTTCTTCGCCGCCG
Phenotype
MIM: 603461 MIM: 605530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC16 PubMed Links

Gene Details

Gene
CDC16
Gene Name
cell division cycle 16
There are no transcripts associated with this gene.

Gene
MIR4502
Gene Name
microRNA 4502
There are no transcripts associated with this gene.

Gene
UPF3A
Gene Name
UPF3 regulator of nonsense transcripts homolog A (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023011.3 338 Silent Mutation CAC,CAT H94H NP_075387.1
NM_080687.2 338 Silent Mutation CAC,CAT H94H NP_542418.1
XM_005266202.4 338 Intron XP_005266259.1
XM_006719991.3 338 Intron XP_006720054.1
XM_011534844.1 338 Intron XP_011533146.1
XM_011534845.2 338 Intron XP_011533147.1
XM_011534846.1 338 Silent Mutation CAC,CAT H94H XP_011533148.1
XM_011534847.2 338 Intron XP_011533149.1
XM_011534848.2 338 Intron XP_011533150.1
XM_017020709.1 338 Intron XP_016876198.1
XM_017020710.1 338 Intron XP_016876199.1
XM_017020711.1 338 Intron XP_016876200.1
XM_017020712.1 338 Silent Mutation CAC,CAT H94H XP_016876201.1

View Full Product Details