Product Details

SNP ID
rs191368819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:149995226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAATTCCATAGGGACGATGCCTG[A/C]CTTCTGCCAACATTTCACCACTGTG
Phenotype
MIM: 606718
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HMGXB3 PubMed Links

Gene Details

Gene
HMGXB3
Gene Name
HMG-box containing 3
There are no transcripts associated with this gene.

Gene
SLC26A2
Gene Name
solute carrier family 26 member 2
There are no transcripts associated with this gene.

Gene
TIGD6
Gene Name
tigger transposable element derived 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243253.1 1898 Missense Mutation GCA,TCA A375S NP_001230182.1
NM_030953.3 1898 Missense Mutation GCA,TCA A375S NP_112215.1

View Full Product Details