Product Details

SNP ID
rs191369801
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124250380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTCGCTCCACTGAGGGCAGGTC[C/G]AAAGCCTTCAGCACTTGTAGCTCCC
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
OR8G1 PubMed Links
Additional Information
For this assay, SNP(s) [rs4482039] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR8G1
Gene Name
olfactory receptor family 8 subfamily G member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002905.1 705 Silent Mutation TCC,TCG S235S NP_001002905.1
Gene
OR8G5
Gene Name
olfactory receptor family 8 subfamily G member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005198.1 705 Intron NP_001005198.1
XM_011542662.2 705 Intron XP_011540964.1
XM_011542663.2 705 Intron XP_011540965.1
XM_011542664.2 705 Intron XP_011540966.1
XM_011542665.2 705 Intron XP_011540967.1
XM_011542666.2 705 Intron XP_011540968.1
XM_011542667.2 705 Intron XP_011540969.1

View Full Product Details