Product Details

SNP ID
rs191738560
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:170532210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTGGGCACTTTTGGGGGTGCC[C/G]GTGGCCCGGGCCGATGGCGCAAGGT
Phenotype
MIM: 607983
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GORAB PubMed Links

Gene Details

Gene
GORAB
Gene Name
golgin, RAB6 interacting
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146039.1 89 Missense Mutation CCG,CGG P21R NP_001139511.1
NM_001320252.1 89 UTR 5 NP_001307181.1
NM_152281.2 89 Missense Mutation CCG,CGG P21R NP_689494.2
XM_006711628.3 89 UTR 5 XP_006711691.1
XM_011510149.2 89 Missense Mutation CCG,CGG P21R XP_011508451.1
XM_011510150.2 89 UTR 5 XP_011508452.1
XM_011510151.1 89 UTR 5 XP_011508453.1
XM_017002807.1 89 UTR 5 XP_016858296.1
Gene
LOC101928650
Gene Name
uncharacterized LOC101928650
There are no transcripts associated with this gene.

View Full Product Details