Product Details

SNP ID
rs191702515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27329150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGCAGTTGTCCTGAGCCACAGT[A/G]ACACCATTGTAGGGAAGCAGCCAGG
Phenotype
MIM: 604883 MIM: 137960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTF3C2 PubMed Links

Gene Details

Gene
GTF3C2
Gene Name
general transcription factor IIIC subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035521.2 2753 Silent Mutation GTC,GTT V670V NP_001030598.1
NM_001318909.1 2753 Silent Mutation GTC,GTT V681V NP_001305838.1
NM_001521.3 2753 Silent Mutation GTC,GTT V670V NP_001512.1
XM_005264272.4 2753 Silent Mutation GTC,GTT V709V XP_005264329.1
XM_011532801.2 2753 Silent Mutation GTC,GTT V785V XP_011531103.1
Gene
GTF3C2-AS1
Gene Name
GTF3C2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MPV17
Gene Name
MPV17, mitochondrial inner membrane protein
There are no transcripts associated with this gene.

View Full Product Details